This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03435862_m1
See other IGF2BP2A GE Assays ›
Gene Symbol:
igf2bp2a
Gene Name:
insulin-like growth factor 2 mRNA binding protein 2a
Gene Aliases:
crfb14, crfb16, igf2bp2, si:ch211-125m20.1
Chromosome Location:
Chr.9: 12467194 - 12576319 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03435862_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: igf2bp2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114558.2 NP_001108030.2 9 - 10 1078 67
XM_005167469.4 XP_005167526.1 9 - 10 1059 67
XM_009304504.3 XP_009302779.1 9 - 10 1828 67
GenBank mRNA BC155087.1 - 1 - 2 63 67
DQ998838.1 - 9 - 10 1078 67
EU296401.2 - 9 - 10 1285 67
FJ418164.1 - 9 - 10 1285 67
FJ418165.1 - 8 - 9 981 67
FJ418166.1 - 8 - 9 981 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
regulation of gene expression
regulation of RNA metabolic process
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0