This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03435095_gH
See other PTPN2A GE Assays ›
Gene Symbol:
ptpn2a
Gene Name:
protein tyrosine phosphatase non-receptor type 2a
Gene Aliases:
PTP-1B, hm:zeh1546, ptpn2, zgc:63623
Chromosome Location:
Chr.16: 10055428 - 10071874 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03435095_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ptpn2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200466.2 NP_956760.2 6 - 7 877 120
GenBank mRNA BC055169.1 - 6 - 7 859 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0