This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434887_m1
See other SH3GL1A GE Assays ›
Gene Symbol:
sh3gl1a
Gene Name:
SH3-domain GRB2-like 1a
Gene Aliases:
sh3gl1b, wu:fd12b05, wu:fy33c10, zgc:56637
Chromosome Location:
Chr.2: 49457631 - 49511458 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434887_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sh3gl1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105606.1 NP_001099076.1 3 - 4 211 90
NM_200301.1 NP_956595.1 3 - 4 329 90
XM_005168418.1 XP_005168475.1 3 - 4 367 90
GenBank mRNA BC049513.1 - 3 - 4 329 90
BC153638.1 - 3 - 4 211 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0