This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434700_g1
See other ARHGAP29B GE Assays ›
Gene Symbol:
arhgap29b
Gene Name:
Rho GTPase activating protein 29b
Gene Aliases:
arhgap29, sb:cb905, zgc:63950
Chromosome Location:
Chr.2: 15128302 - 15197920 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434700_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arhgap29b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200111.2 NP_956405.2 21 - 22 3167 74
XM_005163312.4 XP_005163369.1 21 - 22 3275 74
XM_005163314.4 XP_005163371.1 21 - 22 3258 74
XM_005163315.4 XP_005163372.1 19 - 20 3088 74
GenBank mRNA BC059184.1 - 21 - 22 3160 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
intracellular signal transduction
activation of GTPase activity
GTPase activator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0