This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434688_g1
See other ARHGAP29B GE Assays ›
Gene Symbol:
arhgap29b
Gene Name:
Rho GTPase activating protein 29b
Gene Aliases:
arhgap29, sb:cb905, zgc:63950
Chromosome Location:
Chr.2: 15128302 - 15197920 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434688_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arhgap29b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200111.2 NP_956405.2 10 - 11 1244 64
XM_005163312.4 XP_005163369.1 10 - 11 1352 64
XM_005163314.4 XP_005163371.1 10 - 11 1335 64
XM_005163315.4 XP_005163372.1 8 - 9 1165 64
GenBank mRNA BC059184.1 - 10 - 11 1237 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
intracellular signal transduction
activation of GTPase activity
GTPase activator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0