This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434463_g1
See other EEF1B2 GE Assays ›
Gene Symbol:
eef1b2
Gene Name:
eukaryotic translation elongation factor 1 beta 2
Gene Aliases:
fj06d02, wu:fj06d02, zgc:56277, zgc:86802
Chromosome Location:
Chr.6: 13201010 - 13206255 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434463_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eef1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199949.2 NP_956243.1 3 - 4 612 91
GenBank mRNA AY398339.1 - 3 - 4 612 91
BC071464.1 - 3 - 4 397 91
BC150470.1 - 3 - 4 396 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
nucleic acid binding
translation elongation factor activity
guanyl-nucleotide exchange factor activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0