This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03433386_g1
See other NPC2 GE Assays ›
Gene Symbol:
npc2
Gene Name:
Niemann-Pick disease, type C2
Gene Aliases:
cb292, sb:cb292
Chromosome Location:
Chr.17: 43030320 - 43031809 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03433386_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: npc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173224.1 NP_775331.1 3 - 4 259 85
XM_021467104.1 XP_021322779.1 3 - 4 335 85
GenBank mRNA AF291663.1 - 3 - 4 259 85
BC045895.1 - 3 - 4 256 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid transport
steroid metabolic process
cholesterol metabolic process
sterol transport
intracellular cholesterol transport
cholesterol efflux
cholesterol binding
sterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0