This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432981_g1
See other BAXA GE Assays ›
Gene Symbol:
baxa
Gene Name:
BCL2 associated X, apoptosis regulator a
Gene Aliases:
bax, fj16e01, wu:fc50b10, wu:fj16e01
Chromosome Location:
Chr.3: 37776493 - 37785837 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432981_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: baxa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131562.2 NP_571637.1 5 - 6 500 92
GenBank mRNA BC055592.1 - 5 - 6 500 92
BC164990.1 - 5 - 6 514 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

release of cytochrome c from mitochondria
mitochondrial fusion
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to cadmium ion
response to methylmercury
extrinsic apoptotic signaling pathway in absence of ligand
channel activity
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0