This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432977_m1
See other BAXA GE Assays ›
Gene Symbol:
baxa
Gene Name:
BCL2 associated X, apoptosis regulator a
Gene Aliases:
bax, fj16e01, wu:fc50b10, wu:fj16e01
Chromosome Location:
Chr.3: 37776493 - 37785837 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432977_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: baxa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131562.2 NP_571637.1 1 - 2 54 57
GenBank mRNA BC055592.1 - 1 - 2 54 57
BC164990.1 - 1 - 2 68 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

release of cytochrome c from mitochondria
mitochondrial fusion
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to cadmium ion
response to methylmercury
extrinsic apoptotic signaling pathway in absence of ligand
channel activity
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0