This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432904_m1
See other KDRL GE Assays ›
Gene Symbol:
kdrl
Gene Name:
kinase insert domain receptor like
Gene Aliases:
flk, flk-1, flk1, kdr, kdra, vegfr-2, vegfr2, vegfr2a, vegfr4, vegr2, wu:fk52c05
Chromosome Location:
Chr.14: 32983769 - 33044952 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432904_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: kdrl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131472.1 NP_571547.1 9 - 10 1249 106
XM_009291142.3 XP_009289417.1 9 - 10 1643 106
GenBank mRNA AF180354.1 - 6 - 7 862 106
AF487829.1 - 9 - 10 1599 106
AY056466.1 - 9 - 10 1249 106
BC129158.1 - 9 - 10 1614 106
CU458916.1 - 9 - 10 1614 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
blood vessel development
sprouting angiogenesis
hematopoietic progenitor cell differentiation
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
phosphorylation
cell differentiation
thyroid gland development
positive regulation of kinase activity
embryonic heart tube development
artery morphogenesis
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0