This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432727_m1
See other QKIA GE Assays ›
Gene Symbol:
qkia
Gene Name:
QKI, KH domain containing, RNA binding a
Gene Aliases:
cb344, fb44d06, fc32f06, qk, qk1, qkA, wu:fb44d06, wu:fc32f06, zqk, zqk-1, zqk1
Chromosome Location:
Chr.17: 27723435 - 27813509 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432727_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: qkia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131224.1 NP_571299.1 4 - 5 789 77
XM_009293118.3 XP_009291393.1 4 - 5 811 77
XM_009293119.3 XP_009291394.1 4 - 5 809 77
XM_009293120.3 XP_009291395.1 4 - 5 810 77
GenBank mRNA BC056599.1 - 4 - 5 758 77
BC065667.1 - 4 - 5 757 77
BC155317.1 - 4 - 5 568 77
U62134.1 - 4 - 5 789 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
regulation of translation
multicellular organism development
skeletal muscle tissue development
RNA splicing
regulation of smoothened signaling pathway
skeletal myofibril assembly
cell differentiation
mRNA transport
3'-UTR-mediated mRNA stabilization
nucleic acid binding
RNA binding
mRNA binding
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0