This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432573_g1
See other NDR1 GE Assays ›
Gene Symbol:
ndr1
Gene Name:
nodal-related 1
Gene Aliases:
CHUNP6892, NDR2, Ndr-1, Znr-2, sqt, znr2
Chromosome Location:
Chr.21: 21010919 - 21013085 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432573_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ndr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130966.1 NP_571041.1 1 - 2 325 85
GenBank mRNA AF002218.1 - 1 - 2 453 85
AF056327.1 - 1 - 2 325 85
BC097089.1 - 1 - 2 367 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

determination of left/right asymmetry in lateral mesoderm
mesoderm development
anterior/posterior axis specification, embryo
dorsal/ventral axis specification
dorsal/ventral pattern formation
positive regulation of pathway-restricted SMAD protein phosphorylation
hypothalamus development
BMP signaling pathway
camera-type eye development
positive regulation of fibroblast growth factor receptor signaling pathway
determination of dorsal identity
mesendoderm development
Spemann organizer formation
primitive hemopoiesis
SMAD protein signal transduction
cytokine activity
protein binding
growth factor activity
type I activin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header