This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432228_m1
See other CIP2A GE Assays ›
Gene Symbol:
cip2a
Gene Name:
cellular inhibitor of PP2A
Gene Aliases:
fi40d06, im:7140002, wu:fi40d06, zgc:174160
Chromosome Location:
Chr.24: 25209966 - 25234642 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432228_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cip2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114421.1 NP_001107893.1 9 - 10 1439 71
XM_005162708.4 XP_005162765.1 9 - 10 1488 71
XM_021470191.1 XP_021325866.1 10 - 11 1178 71
GenBank mRNA BC155117.1 - 9 - 10 1439 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0