This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432092_m1
See other ABLIM1A GE Assays ›
Gene Symbol:
ablim1a
Gene Name:
actin binding LIM protein 1a
Gene Aliases:
ablim1, zgc:172321
Chromosome Location:
Chr.13: 43507383 - 43599920 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432092_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ablim1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114316.1 NP_001107788.1 2 - 3 257 67
XM_009307552.3 XP_009305827.1 2 - 3 2692 67
XM_009307554.3 XP_009305829.1 2 - 3 2692 67
XM_009307559.3 XP_009305834.1 2 - 3 2692 67
XM_009307565.3 XP_009305840.1 2 - 3 2692 67
XM_017358665.2 XP_017214154.1 2 - 3 2692 67
XM_017358666.2 XP_017214155.1 2 - 3 2692 67
XM_017358667.2 XP_017214156.1 2 - 3 2691 67
XM_021480476.1 XP_021336151.1 2 - 3 2693 67
XM_021480477.1 XP_021336152.1 2 - 3 2693 67
XM_021480478.1 XP_021336153.1 2 - 3 2692 67
XM_021480479.1 XP_021336154.1 2 - 3 2692 67
XM_021480480.1 XP_021336155.1 2 - 3 2692 67
XM_021480481.1 XP_021336156.1 2 - 3 2692 67
XM_021480482.1 XP_021336157.1 2 - 3 2692 67
XM_021480483.1 XP_021336158.1 2 - 3 2692 67
GenBank mRNA BC155652.1 - 2 - 3 257 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
lamellipodium assembly
cilium morphogenesis
actin binding
metal ion binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0