This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03431107_m1
See other MBNL3 GE Assays ›
Gene Symbol:
mbnl3
Gene Name:
muscleblind-like splicing regulator 3
Gene Aliases:
mbnl3a, mbnl3b, mbnl3c
Chromosome Location:
Chr.14: 30986754 - 31080162 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03431107_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mbnl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001112675.1 NP_001106145.1 1 - 2 245 66
XM_009291127.3 XP_009289402.1 3 - 4 1643 66
XM_009291128.3 XP_009289403.1 2 - 3 1411 66
XM_009291129.3 XP_009289404.1 2 - 3 1293 66
XM_021481047.1 XP_021336722.1 2 - 3 1411 66
XM_021481048.1 XP_021336723.1 2 - 3 1408 66
XM_021481049.1 XP_021336724.1 2 - 3 1410 66
XM_021481050.1 XP_021336725.1 2 - 3 1408 66
XM_021481051.1 XP_021336726.1 2 - 3 1408 66
XM_021481052.1 XP_021336727.1 2 - 3 1409 66
XM_021481053.1 XP_021336728.1 2 - 3 1409 66
GenBank mRNA EU284561.1 - 1 - 2 245 66
EU284562.1 - 1 - 2 245 66
EU284563.1 - 1 - 2 245 66
EU284564.1 - 1 - 2 245 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0