This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03430790_m1
See other ABCD1 GE Assays ›
Gene Symbol:
abcd1
Gene Name:
ATP-binding cassette, sub-family D (ALD), member 1
Gene Aliases:
ALDP, zgc:172102
Chromosome Location:
Chr.8: 8045058 - 8085429 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03430790_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: abcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111186.1 NP_001104656.1 5 - 6 1526 63
XM_005166714.4 XP_005166771.1 5 - 6 2554 63
GenBank mRNA BC155605.1 - 5 - 6 1526 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
fatty acid beta-oxidation
peroxisome organization
oligodendrocyte development
peroxisomal long-chain fatty acid import
cholesterol transport
adrenal gland development
positive regulation of myelination
swimming behavior
very long-chain fatty acid catabolic process
transmembrane transport
motor behavior
nucleotide binding
long-chain fatty acid transporter activity
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0