This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03430712_m1
See other ETF1A GE Assays ›
Gene Symbol:
etf1a
Gene Name:
eukaryotic translation termination factor 1a
Gene Aliases:
zgc:171591
Chromosome Location:
Chr.14: 2589460 - 2616596 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03430712_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: etf1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111171.1 NP_001104641.1 8 - 9 1219 84
GenBank mRNA BC153983.1 - 8 - 9 1219 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation release factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translational termination
translational termination
translation release factor activity
translation release factor activity, codon specific
sequence-specific mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0