This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03429284_m1
See other DDX3XA GE Assays ›
Gene Symbol:
ddx3xa
Gene Name:
DEAD-box helicase 3 X-linked a
Gene Aliases:
ddx3, ddx3a, fb74g09, wu:fb74g09, zgc:158804
Chromosome Location:
Chr.9: 33334456 - 33354886 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03429284_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ddx3xa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001126423.1 NP_001119895.1 3 - 4 440 93
XM_009304751.3 XP_009303026.1 3 - 4 484 93
GenBank mRNA BC133162.1 - 3 - 4 440 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

gamete generation
cell differentiation
neutrophil chemotaxis
nucleotide binding
nucleic acid binding
RNA binding
RNA helicase activity
helicase activity
ATP binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header