This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03428663_g1
See other APOA4B.1 GE Assays ›
Gene Symbol:
apoa4b.1 apoa4b.2
Gene Name:
apolipoprotein A-IV b, tandem duplicate 1
apolipoprotein A-IV b, tandem duplicate 2
Gene Aliases:
apoa4, wu:fb64f03, wu:fb66f09, zgc:122983
fc28d04, wu:fc10a03, wu:fc28d04, zgc:194131
Chromosome Location:
Chr.16: 23914078 - 23915980 on Build GRCz11
Chromosome Location:
Chr.16: 23947078 - 23948538 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
-
Assay ID Dr03428663_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: apoa4b.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079861.1 NP_001073330.1 3 - 4 247 85
GenBank mRNA BC076032.1 - 2 - 3 207 85
BC093239.1 - 3 - 4 230 85
BC107620.1 - 3 - 4 247 85
Gene Symbol: apoa4b.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128758.1 NP_001122230.1 3 - 4 239 85
XM_021466732.1 XP_021322407.1 3 - 4 380 85
XM_021466733.1 XP_021322408.1 3 - 4 275 85
GenBank mRNA BC162715.1 - 3 - 4 239 85
BC162716.1 - 3 - 4 239 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

apolipoprotein

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
lipoprotein metabolic process
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0