This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03428463_m1
See other SMPD5 GE Assays ›
Gene Symbol:
smpd5
Gene Name:
sphingomyelin phosphodiesterase 5
Gene Aliases:
mtnSMase, smpdm, zgc:153995
Chromosome Location:
Chr.16: 53518889 - 53542242 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03428463_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: smpd5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077615.1 NP_001071083.1 2 - 3 1189 80
XM_005158452.4 XP_005158509.1 2 - 3 1416 80
XM_005158453.4 XP_005158510.1 2 - 3 1428 80
GenBank mRNA AB361066.1 - 1 - 2 1020 80
BC124788.1 - 2 - 3 1189 80
BC155730.1 - 2 - 3 1201 80
BC165400.1 - 1 - 2 1054 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
sphingomyelin metabolic process
glycosphingolipid metabolic process
phospholipase activity
sphingomyelin phosphodiesterase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0