This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03427259_m1
See other GRID2IPB GE Assays ›
Gene Symbol:
grid2ipb
Gene Name:
glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein, b
Gene Aliases:
grid2ip, si:ch211-147h1.3, zC147H1.3
Chromosome Location:
Chr.12: 574217 - 626780 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03427259_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: grid2ipb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044903.1 NP_001038368.1 9 - 10 2105 100
XM_009306633.3 XP_009304908.1 16 - 17 3503 100
XM_017358463.2 XP_017213952.2 16 - 17 3491 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Danio rerio Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0