This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03426178_m1
See other SMPD2A GE Assays ›
Gene Symbol:
smpd2a
Gene Name:
sphingomyelin phosphodiesterase 2a
Gene Aliases:
NSMase, smpd2
Chromosome Location:
Chr.23: 5565736 - 5588492 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03426178_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: smpd2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030219.2 NP_001025390.2 3 - 4 243 60
XM_005170835.4 XP_005170892.1 4 - 5 499 60
GenBank mRNA AB196165.1 - 3 - 4 223 60
BC163237.1 - 3 - 4 250 60
EH590607.1 - 3 - 4 252 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
sphingomyelin metabolic process
sphingolipid catabolic process
ceramide biosynthetic process
cellular response to xenobiotic stimulus
cellular response to temperature stimulus
sphingolipid mediated signaling pathway
phospholipase activity
sphingomyelin phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0