This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03425842_m1
See other GOLGA7 GE Assays ›
Gene Symbol:
golga7
Gene Name:
golgin A7
Gene Aliases:
wu:fc23b09, zgc:86753
Chromosome Location:
Chr.10: 20364388 - 20372146 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03425842_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: golga7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105599.1 NP_001099069.1 2 - 3 168 91
XM_021479212.1 XP_021334887.1 2 - 3 218 91
GenBank mRNA BC071425.1 - 2 - 3 167 91
BC153595.1 - 2 - 3 168 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
peptidyl-L-cysteine S-palmitoylation
Golgi to plasma membrane protein transport
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0