This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03424683_m1
See other SLC5A6B GE Assays ›
Gene Symbol:
slc5a6b
Gene Name:
solute carrier family 5 member 6
Gene Aliases:
slc5a6, wu:fa19f06, zgc:114067
Chromosome Location:
Chr.17: 6535988 - 6556143 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03424683_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc5a6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025528.2 NP_001020699.1 5 - 6 757 102
XM_005158575.4 XP_005158632.1 5 - 6 964 102
XM_005158576.4 XP_005158633.1 5 - 6 964 102
GenBank mRNA BC097129.1 - 5 - 6 758 102
BC155140.1 - 5 - 6 763 102
BC171549.1 - 4 - 5 585 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
biotin transport
pantothenate transmembrane transport
transmembrane transport
sodium-dependent multivitamin transmembrane transporter activity
symporter activity
transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0