This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03423229_g1
See other AGR2 GE Assays ›
Gene Symbol:
agr2
Gene Name:
anterior gradient 2
Gene Aliases:
sb:eu228, wu:fj29g05, zgc:112187
Chromosome Location:
Chr.19: 30401547 - 30404099 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03423229_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: agr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012481.2 NP_001012499.1 7 - 8 540 61
GenBank mRNA AY796218.1 - 7 - 8 536 61
BC093250.1 - 7 - 8 537 61
BC152144.1 - 7 - 8 539 61
EH453285.1 - 7 - 8 563 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell-substrate adhesion
negative regulation of cell death
intestinal epithelial cell differentiation
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0