This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03423099_m1
See other UBE2KB GE Assays ›
Gene Symbol:
ube2kb
Gene Name:
ubiquitin-conjugating enzyme E2Kb (UBC1 homolog, yeast)
Gene Aliases:
hip2, ube2k, zgc:103472
Chromosome Location:
Chr.1: 47087 - 57270 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03423099_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ube2kb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008611.2 NP_001008611.1 5 - 6 855 84
GenBank mRNA BC086816.1 - 5 - 6 856 84
BC154174.1 - 5 - 6 567 84
BC165002.1 - 5 - 6 434 84
EH554183.1 - 5 - 6 716 84
EH575297.1 - 2 - 3 652 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
nucleotide binding
ATP binding
transferase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0