This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03190431_s1
See other B2ML GE Assays ›
Gene Symbol:
b2ml
Gene Name:
beta-2-microglobulin, like
Gene Aliases:
b2m, wu:fb66f11, wu:fj01f08, zgc:64161
Chromosome Location:
Chr.8: 50942240 - 50950549 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03190431_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: b2ml
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213126.2 NP_998291.2 4 795 139
GenBank mRNA BC057435.1 - 752 139
EB935339.1 - 1 92 139

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

immune system process
antigen processing and presentation of peptide antigen via MHC class I
immune response

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0