Assay Details
| Gene Symbol: asic1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214791.1 | NP_999956.1 | 9 - 10 | 1668 | 88 | |
| XM_005161666.4 | XP_005161723.1 | 9 - 10 | 1952 | 88 | ||
| XM_005161667.4 | XP_005161724.1 | 10 - 11 | 2404 | 88 | ||
| GenBank mRNA | AJ609615.1 | - | 9 - 10 | 1668 | 88 | |
| BC162362.1 | - | 9 - 10 | 1637 | 88 | ||
| BC162368.1 | - | 9 - 10 | 1637 | 88 | ||
More Information
Set Membership: |
|

asic1b
Genomic Map