This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03152547_m1
See other ASIC1B GE Assays ›
Gene Symbol:
asic1b
Gene Name:
acid-sensing (proton-gated) ion channel 1b
Gene Aliases:
ASIC1, ASIC1-B, accn2a, zASIC1.1
Chromosome Location:
Chr.22: 7042822 - 7401401 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03152547_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: asic1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214791.1 NP_999956.1 7 - 8 1484 72
XM_005161666.4 XP_005161723.1 7 - 8 1768 72
XM_005161667.4 XP_005161724.1 8 - 9 2220 72
GenBank mRNA AJ609615.1 - 7 - 8 1484 72
BC162362.1 - 7 - 8 1453 72
BC162368.1 - 7 - 8 1453 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

ion transport
sodium ion transport
sodium ion transmembrane transport
protein homotrimerization
cellular response to pH
sodium channel activity
ligand-gated sodium channel activity
acid-sensing ion channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0