This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03151329_g1
See other ST3GAL3A GE Assays ›
Gene Symbol:
st3gal3a
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3a
Gene Aliases:
siat6, st3ga3-r, st3gal3, zgc:63978
Chromosome Location:
Chr.6: 3334358 - 3405499 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03151329_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: st3gal3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200355.1 NP_956649.1 7 - 8 596 137
XM_021476802.1 XP_021332477.1 7 - 8 689 137
XM_021476804.1 XP_021332479.1 7 - 8 919 137
XM_021476805.1 XP_021332480.1 6 - 7 607 137
XM_021476806.1 XP_021332481.1 6 - 7 755 137
XM_021476807.1 XP_021332482.1 8 - 9 743 137
GenBank mRNA AJ626820.1 - 7 - 8 623 137
AJ705063.1 - 6 - 7 444 137
BC053179.1 - 7 - 8 596 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
sialyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0