This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03151009_g1
See other NAPAB GE Assays ›
Gene Symbol:
napab
Gene Name:
N-ethylmaleimide-sensitive factor attachment protein, alpha b
Gene Aliases:
napa, wu:fa26c04, zgc:63906
Chromosome Location:
Chr.18: 44673996 - 44696810 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03151009_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: napab
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199766.1 NP_956060.1 8 - 9 734 88
GenBank mRNA AY398385.1 - 8 - 9 740 88
BC054568.1 - 8 - 9 734 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
regulation of synaptic vesicle priming
protein transport
vesicle-mediated transport
synaptic transmission, glutamatergic
SNARE complex disassembly
soluble NSF attachment protein activity
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0