This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03150413_m1
See other CD74A GE Assays ›
Gene Symbol:
cd74a
Gene Name:
CD74 molecule, major histocompatibility complex, class II invariant chain a
Gene Aliases:
Iclp-1, iclp1, wu:fb11f09, wu:fb12a01, wu:fb13d06, wu:fk94e07
Chromosome Location:
Chr.14: 3038597 - 3052361 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03150413_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cd74a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131590.1 NP_571665.1 4 - 5 466 109
GenBank mRNA AF114830.1 - 4 - 5 463 109
AF148214.1 - 4 - 5 466 109
BC065433.1 - 4 - 5 456 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine-mediated signaling pathway
T cell activation involved in immune response
dendritic cell antigen processing and presentation
positive regulation of type 2 immune response
intracellular protein transport
immune response
antigen processing and presentation
positive regulation of kinase activity
negative regulation of DNA damage response, signal transduction by p53 class mediator
inner ear development
neuromast development
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of macrophage cytokine production
positive regulation of ERK1 and ERK2 cascade
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
cytokine receptor activity
macrophage migration inhibitory factor binding
MHC class II protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0