This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03149940_m1
See other TRIOA GE Assays ›
Gene Symbol:
trioa
Gene Name:
trio Rho guanine nucleotide exchange factor a
Gene Aliases:
si:ch211-13c6.1, si:dkey-158b13.2, trio
Chromosome Location:
Chr.19: 41840964 - 41991299 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03149940_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: trioa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001104526.1 NP_001097996.1 31 - 32 4748 70
XM_009294421.3 XP_009292696.1 31 - 32 4915 70
XM_017352243.2 XP_017207732.1 31 - 32 4919 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
axon guidance
central nervous system development
phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0