This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03148776_m1
See other RAD21B GE Assays ›
Gene Symbol:
rad21b
Gene Name:
RAD21 cohesin complex component b
Gene Aliases:
si:ch211-246f9.1
Chromosome Location:
Chr.19: 44067618 - 44091405 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03148776_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rad21b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045120.2 NP_001038585.1 12 - 13 1720 182
XM_005159782.4 XP_005159839.1 13 - 14 1872 182

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sister chromatid cohesion
mitotic sister chromatid cohesion
synaptonemal complex assembly
meiotic sister chromatid cohesion
replication-born double-strand break repair via sister chromatid exchange
chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0