This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03148494_m1
See other SLC6A4A GE Assays ›
Gene Symbol:
slc6a4a
Gene Name:
solute carrier family 6 member 4a
Gene Aliases:
-
Chromosome Location:
Chr.15: 28607631 - 28634544 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03148494_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc6a4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039972.1 NP_001035061.1 12 - 13 1949 115
XM_005157654.4 XP_005157711.1 15 - 16 3127 115
XM_005157655.4 XP_005157712.1 15 - 16 3050 115
XM_009291668.3 XP_009289943.1 16 - 17 3319 115
XM_009291669.3 XP_009289944.1 16 - 17 3237 115
XM_009291670.3 XP_009289945.1 14 - 15 2858 115
XM_009291671.3 XP_009289946.1 16 - 17 2615 115
XM_009291672.3 XP_009289947.1 14 - 15 2776 115
XM_009291673.3 XP_009289948.1 12 - 13 1698 115
GenBank mRNA BC163766.1 - 12 - 13 1949 115
BC163777.1 - 12 - 13 1949 115
DQ285098.1 - 12 - 13 1949 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
serotonin transport
sodium ion transmembrane transport
serotonin uptake
neurotransmitter:sodium symporter activity
serotonin:sodium symporter activity
symporter activity
serotonin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0