Assay Details
| Gene Symbol: cox7a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032728.1 | NP_001027900.1 | 2 - 3 | 184 | 72 | |
| GenBank mRNA | BC095093.1 | - | 2 - 3 | 184 | 72 | |
More Information
Set Membership: |
|

cox7a2a
Genomic Map