This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03146942_m1
See other USB1 GE Assays ›
Gene Symbol:
usb1
Gene Name:
U6 snRNA biogenesis 1
Gene Aliases:
zgc:91896
Chromosome Location:
Chr.25: 13677289 - 13686754 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03146942_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: usb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003460.1 NP_001003460.1 2 - 3 415 77
XM_009297898.3 XP_009296173.1 2 - 3 478 77
XM_009297899.2 XP_009296174.1 2 - 3 340 77
XM_021470478.1 XP_021326153.1 2 - 3 342 77
GenBank mRNA BC077163.1 - 2 - 3 415 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

hemopoiesis
neutrophil differentiation
U6 snRNA 3'-end processing
regulation of RNA splicing
3'-5'-exoribonuclease activity
nuclease activity
hydrolase activity
poly(U)-specific exoribonuclease activity, producing 3' uridine cyclic phosphate ends

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0