This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03146918_m1
See other PRCP GE Assays ›
Gene Symbol:
prcp
Gene Name:
prolylcarboxypeptidase (angiotensinase C)
Gene Aliases:
zgc:91816
Chromosome Location:
Chr.15: 11218246 - 11303889 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03146918_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: prcp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002694.1 NP_001002694.1 3 - 4 621 80
XM_009291512.3 XP_009289787.1 3 - 4 814 80
XM_009291513.3 XP_009289788.1 3 - 4 819 80
XM_017359231.2 XP_017214720.1 3 - 4 370 80
XM_021481379.1 XP_021337054.1 3 - 4 817 80
XM_021481380.1 XP_021337055.1 3 - 4 816 80
GenBank mRNA BC076507.1 - 3 - 4 621 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of systemic arterial blood pressure
proteolysis
regulation of blood vessel endothelial cell migration
angiogenesis involved in wound healing
carboxypeptidase activity
peptidase activity
serine-type peptidase activity
dipeptidyl-peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0