This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03146807_m1
See other ARL1 GE Assays ›
Gene Symbol:
arl1
Gene Name:
ADP-ribosylation factor-like 1
Gene Aliases:
zgc:92883
Chromosome Location:
Chr.4: 5910255 - 5913412 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03146807_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002473.1 NP_001002473.1 4 - 5 407 88
GenBank mRNA BC076341.1 - 4 - 5 407 88
BC164960.1 - 3 - 4 367 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
Golgi organization
toxin metabolic process
vesicle-mediated transport
activation of phospholipase D activity
protein localization to Golgi apparatus
retrograde transport, endosome to Golgi
nucleotide binding
GTPase activity
GTP binding
enzyme activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0