This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03145582_m1
See other ARHGAP12B GE Assays ›
Gene Symbol:
arhgap12b
Gene Name:
Rho GTPase activating protein 12b
Gene Aliases:
arhgap12, zgc:73239
Chromosome Location:
Chr.12: 75425 - 182061 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03145582_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arhgap12b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200874.1 NP_957168.1 14 - 15 2302 81
XM_009306595.3 XP_009304870.1 15 - 16 2411 81
XM_021480040.1 XP_021335715.1 16 - 17 2534 81
XM_021480041.1 XP_021335716.1 16 - 17 2531 81
XM_021480042.1 XP_021335717.1 15 - 16 2462 81
XM_021480043.1 XP_021335718.1 15 - 16 1854 81
GenBank mRNA BC063321.1 - 14 - 15 2302 81
BC065619.1 - 15 - 16 3104 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of GTPase activity
GTPase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0