This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03145510_m1
See other GNAO1B GE Assays ›
Gene Symbol:
gnao1b
Gene Name:
guanine nucleotide binding protein (G protein), alpha activating activity polypeptide O, b
Gene Aliases:
zgc:73153
Chromosome Location:
Chr.7: 18081 - 29123 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03145510_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gnao1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200828.1 NP_957122.1 3 - 4 377 87
GenBank mRNA BC060909.1 - 3 - 4 377 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
dopamine receptor signaling pathway
nucleotide binding
G-protein coupled receptor binding
GTPase activity
GTP binding
guanyl nucleotide binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
mu-type opioid receptor binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0