This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03144270_m1
See other ACVR1L GE Assays ›
Gene Symbol:
acvr1l
Gene Name:
activin A receptor, type 1 like
Gene Aliases:
acvr1, alk2, alk8, alk8sum, etID124543.2, fk50h05, laf, lost-a-fin, mfn, wu:fk50h05
Chromosome Location:
Chr.2: 41524235 - 41543760 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03144270_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: acvr1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131345.1 NP_571420.1 3 - 4 257 68
XM_021478751.1 XP_021334426.1 2 - 3 210 68
GenBank mRNA AF038425.1 - 3 - 4 257 68
BC162313.1 - 2 - 3 122 68
BC162317.1 - 2 - 3 122 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
liver development
vasculature development
heart looping
determination of left/right asymmetry in lateral mesoderm
embryonic heart tube morphogenesis
heart jogging
cardiac atrium development
proepicardium development
protein phosphorylation
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
determination of left/right symmetry
brain development
heart development
anterior/posterior pattern specification
dorsal/ventral pattern formation
phosphorylation
central nervous system formation
BMP signaling pathway
regulation of BMP signaling pathway
embryonic heart tube development
embryonic caudal fin morphogenesis
embryonic hemopoiesis
determination of left/right asymmetry in diencephalon
odontogenesis
negative regulation of calcium-dependent cell-cell adhesion
determination of ventral identity
mesoderm morphogenesis
lateral mesoderm development
cartilage development
dorsal convergence
determination of heart left/right asymmetry
cellular response to growth factor stimulus
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
protein binding
ATP binding
kinase activity
transferase activity
SMAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0