This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03143331_g1
See other CACNB2B GE Assays ›
Gene Symbol:
cacnb2b
Gene Name:
calcium channel, voltage-dependent, beta 2b
Gene Aliases:
CACNB2.2, cacnb2a, im:7141271, si:dkey-32m20.2
Chromosome Location:
Chr.2: 31462762 - 31539459 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03143331_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cacnb2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081748.2 NP_001075217.2 763 75
NM_001166324.1 NP_001159796.1 521 75
GenBank mRNA BC160620.1 - 4 - 5 412 75
BC162962.1 - 4 - 5 379 75
BC162994.1 - 4 - 5 379 75
DQ372944.1 - 4 - 5 412 75
EU301442.1 - 763 75
EU301443.1 - 521 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chemical synaptic transmission
neuromuscular junction development
calcium ion transmembrane transport
regulation of voltage-gated calcium channel activity
voltage-gated calcium channel activity
high voltage-gated calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0