This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03143278_g1
See other DNAI1.2 GE Assays ›
Gene Symbol:
dnai1.2
Gene Name:
dynein, axonemal, intermediate chain 1, paralog 2
Gene Aliases:
zgc:158666
Chromosome Location:
Chr.21: 17956062 - 18049419 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03143278_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnai1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080558.1 NP_001074027.1 4 - 5 367 108
XM_009295313.3 XP_009293588.1 4 - 5 395 108
XM_009295314.3 XP_009293589.1 4 - 5 395 108
XM_021468848.1 XP_021324523.1 4 - 5 264 108
GenBank mRNA BC129365.1 - 4 - 5 367 108
BC164796.1 - 4 - 5 319 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

microtubule or microtubule-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cilium movement
microtubule-based movement
outer dynein arm assembly
dynein light chain binding
dynein heavy chain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0