Assay Details
Gene Symbol: dnai1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080558.1 | NP_001074027.1 | 3 - 4 | 290 | 79 | |
GenBank mRNA | BC129365.1 | - | 3 - 4 | 290 | 79 | |
BC164796.1 | - | 3 - 4 | 242 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
