This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03141924_m1
See other PER1A GE Assays ›
Gene Symbol:
per1a
Gene Name:
period circadian clock 1a
Gene Aliases:
per1, si:ch211-237l4.4, zC237L4.4
Chromosome Location:
Chr.10: 22811619 - 22831513 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03141924_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: per1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030183.1 NP_001025354.1 5 - 6 967 83
XM_005172625.4 XP_005172682.1 6 - 7 1378 83
XM_005172626.4 XP_005172683.1 6 - 7 1378 83
XM_005172627.4 XP_005172684.1 6 - 7 1378 83
XM_021479187.1 XP_021334862.1 6 - 7 1378 83
GenBank mRNA BC163543.1 - 5 - 6 1036 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to light stimulus
photoperiodism
circadian regulation of gene expression
response to hydrogen peroxide
entrainment of circadian clock by photoperiod
transcription regulatory region sequence-specific DNA binding
transcription corepressor binding
transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0