This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03141850_m1
See other OTOFA GE Assays ›
Gene Symbol:
otofa
Gene Name:
otoferlin a
Gene Aliases:
fj34b10, otof, si:dkey-181f18.3, wu:fj34b10
Chromosome Location:
Chr.20: 31076238 - 31212903 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03141850_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: otofa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030112.2 NP_001025283.1 34 - 35 4209 74
XM_009294732.3 XP_009293007.1 35 - 36 4664 74
XM_009294733.3 XP_009293008.1 35 - 36 4665 74
XM_009294734.3 XP_009293009.1 35 - 36 4664 74
XM_009294736.3 XP_009293011.1 35 - 36 4664 74
XM_009294738.3 XP_009293013.1 35 - 36 4635 74
XM_009294739.3 XP_009293014.1 34 - 35 4592 74
XM_009294740.3 XP_009293015.1 35 - 36 4580 74
XM_009294742.3 XP_009293017.1 34 - 35 4508 74
XM_009294744.3 XP_009293019.1 34 - 35 4508 74
XM_021468441.1 XP_021324116.1 34 - 35 4634 74
XM_021468442.1 XP_021324117.1 34 - 35 4550 74
XM_021468443.1 XP_021324118.1 34 - 35 4592 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

startle response
plasma membrane organization
sensory perception of sound
response to auditory stimulus
synaptic vesicle exocytosis
synaptic vesicle priming
neuromuscular process controlling balance
calcium ion binding
AP-2 adaptor complex binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0