This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03141514_m1
See other DNMT3AB GE Assays ›
Gene Symbol:
dnmt3ab
Gene Name:
DNA (cytosine-5-)-methyltransferase 3 alpha b
Gene Aliases:
dnmt3a1, dnmt6
Chromosome Location:
Chr.17: 36937808 - 37054963 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03141514_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnmt3ab
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001018140.2 NP_001018150.1 8 - 9 1321 83
NM_001328167.1 NP_001315096.1 13 - 14 1870 83
XM_005158845.4 XP_005158902.1 8 - 9 1235 83
XM_009293199.3 XP_009291474.1 12 - 13 1757 83
XM_009293200.3 XP_009291475.1 8 - 9 1328 83
XM_021467161.1 XP_021322836.1 13 - 14 2010 83
XM_021467162.1 XP_021322837.1 13 - 14 1920 83
XM_021467163.1 XP_021322838.1 13 - 14 1921 83
GenBank mRNA AB196917.1 - 8 - 9 1323 83
BC162582.1 - 8 - 9 1233 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
regulation of gene expression
methylation
methyltransferase activity
DNA-methyltransferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0