Assay Details
Gene Symbol: fn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013261.1 | NP_001013279.1 | 39 - 40 | 6595 | 135 | |
XM_005160284.3 | XP_005160341.1 | 39 - 40 | 6627 | 135 | ||
XM_009305554.3 | XP_009303829.1 | 40 - 41 | 6907 | 135 | ||
GenBank mRNA | AF500198.1 | - | 1 - 2 | 372 | 135 | |
AY193829.1 | - | 1 - 2 | 123 | 135 | ||
AY538257.1 | - | 39 - 40 | 6595 | 135 | ||
AY725818.1 | - | 40 - 41 | 6886 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
