Assay Details
Gene Symbol: fn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013261.1 | NP_001013279.1 | 13 - 14 | 2026 | 90 | |
XM_005160284.3 | XP_005160341.1 | 13 - 14 | 2061 | 90 | ||
XM_009305554.3 | XP_009303829.1 | 13 - 14 | 2062 | 90 | ||
GenBank mRNA | AY538257.1 | - | 13 - 14 | 2026 | 90 | |
AY725818.1 | - | 13 - 14 | 2041 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
